ENA371178192 Enamine

N-methyl-2-(N-methylacetamido)acetamide

  • MDL number MFCD19595954
  • Cas Number : 24131-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108323
Availability details for
Quantity
  • Availability
  • Add to Cart