ENA371178194 Enamine

methyl 3-{[(pyridin-3-yl)methyl]amino}propanoate

  • MDL number MFCD10687257
  • Cas Number : 33710-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106381
Availability details for
Quantity
  • Availability
  • Add to Cart