ENA371178197 Enamine

N-(2,2,2-trifluoroethyl)-4-(trifluoromethyl)aniline

  • MDL number MFCD11152022
  • Cas Number : 1021128-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-108335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108335
Availability details for
Quantity
  • Availability
  • Add to Cart