ENA371178199 Enamine

3-(chloromethyl)-N,N-dimethyl-1,2,4-thiadiazol-5-amine

  • MDL number MFCD20623535
  • Cas Number : 32577-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106398
Availability details for
Quantity
  • Availability
  • Add to Cart