ENA371178210 Enamine

2,5-difluoro-4-methoxyaniline

  • MDL number MFCD18394891
  • Cas Number : 394223-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-106400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106400
Availability details for
Quantity
  • Availability
  • Add to Cart