ENA371178211 Enamine

1-{thieno[3,2-b]thiophen-2-yl}methanamine

  • MDL number MFCD08059492
  • Cas Number : 874881-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108353
Availability details for
Quantity
  • Availability
  • Add to Cart