ENA371178218 Enamine

N-({imidazo[1,2-a]pyridin-3-yl}methylidene)hydroxylamine

  • Cas Number : 30493-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106404
Availability details for
Quantity
  • Availability
  • Add to Cart