ENA371178220 Enamine

4-(tert-butoxy)butanenitrile

  • MDL number MFCD11196944
  • Cas Number : 33563-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102437
Availability details for
Quantity
  • Availability
  • Add to Cart