ENA371178221 Enamine

2-(5-fluoro-2-nitrophenoxy)-2-phenylacetic acid

  • MDL number MFCD09802940
  • Cas Number : 1016504-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108367
Availability details for
Quantity
  • Availability
  • Add to Cart