ENA371178224 Enamine

N-[4-chloro-3-(trifluoromethyl)phenyl]-2-hydroxybenzamide

  • MDL number MFCD01428896
  • Cas Number : 369-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108395
Availability details for
Quantity
  • Availability
  • Add to Cart