ENA371178227 Enamine

2-hydroxy-N-[3-(trifluoromethyl)phenyl]benzamide

  • MDL number MFCD00868387
  • Cas Number : 587-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-186
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108398
Availability details for
Quantity
  • Availability
  • Add to Cart