ENA371178238 Enamine

2,2-diethylbutanoic acid

  • MDL number MFCD00053496
  • Cas Number : 813-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108409

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108409
Availability details for
Quantity
  • Availability
  • Add to Cart