ENA371178246 Enamine

2-acetamido-N,N-dimethylpropanamide

  • MDL number MFCD20355180
  • Cas Number : 24847-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108434
Availability details for
Quantity
  • Availability
  • Add to Cart