ENA371178247 Enamine

4-(aminomethyl)-1-(propan-2-yl)piperidin-4-ol

  • MDL number MFCD12169415
  • Cas Number : 132740-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108435
Availability details for
Quantity
  • Availability
  • Add to Cart