ENA371178248 Enamine

2-[(5-amino-1H-1,2,4-triazol-3-yl)sulfanyl]ethan-1-ol

  • MDL number MFCD08273687
  • Cas Number : 941867-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126...128
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102533

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102533
Availability details for
Quantity
  • Availability
  • Add to Cart