ENA371178257 Enamine

O,O-dimethyl (methylsulfanyl)phosphonothioate

  • MDL number MFCD01705166
  • Cas Number : 2953-29-9
  • Molecular Weight : 172.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name O,O,S-trimethyl dithiophosphate
Inchi Code 1S/C3H9O2PS2/c1-4-6(7,5-2)8-3/h1-3H3
Inchi Key ILDHFYGNYDICFB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-108463

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P273,P280,P301+P310,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P391,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-108463
pictograms GHS06,GHS09
hstatements H301,H312,H315,H319,H332,H335,H411
Availability details for
Quantity
  • Availability
  • Add to Cart