ENA371178260 Enamine

4-(3,4-dimethoxybenzamido)-2-hydroxybenzoic acid

  • MDL number MFCD13878431
  • Cas Number : 1030572-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108504
Availability details for
Quantity
  • Availability
  • Add to Cart