ENA371178263 Enamine

methyl 2-methanesulfinylacetate

  • MDL number MFCD00002092
  • Cas Number : 52147-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108513

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-108513
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart