ENA371178265 Enamine

methyl 4-hydroxyoxane-4-carboxylate

  • MDL number MFCD12031301
  • Cas Number : 115996-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108519
Availability details for
Quantity
  • Availability
  • Add to Cart