ENA371178277 Enamine

2-[(1,3,5-trimethyl-1H-pyrazol-4-yl)oxy]acetic acid

  • MDL number MFCD20825190
  • Cas Number : 1384713-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106452

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106452
Availability details for
Quantity
  • Availability
  • Add to Cart