ENA371178302 Enamine

1-amino-4-ethylcyclohexane-1-carboxylic acid

  • MDL number MFCD07776231
  • Cas Number : 69164-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39473
Availability details for
Quantity
  • Availability
  • Add to Cart