ENA371178303 Enamine

4-(bromomethyl)-1-chloro-2-nitrobenzene

  • MDL number MFCD12025138
  • Cas Number : 3360-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39674

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-39674
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart