ENA371178311 Enamine

2-chloro-N-[2-chloro-5-(1H-1,2,3,4-tetrazol-1-yl)phenyl]acetamide

  • MDL number MFCD12123935
  • Cas Number : 1153017-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47690

Safety Information

msds https://www.enaminestore.com/catalog/EN300-47690
Availability details for
Quantity
  • Availability
  • Add to Cart