ENA371178326 Enamine

3-chloropyrazine-2-carbaldehyde

  • MDL number MFCD07371660
  • Cas Number : 121246-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60212

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60212
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart