ENA371178344 Enamine

N-(prop-2-yn-1-yl)acetamide

  • MDL number MFCD14631295
  • Cas Number : 65881-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-78
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76249
Availability details for
Quantity
  • Availability
  • Add to Cart