ENA371178353 Enamine

2-amino-N-(2-hydroxyethyl)acetamide

  • MDL number MFCD11650491
  • Cas Number : 75007-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82181
Availability details for
Quantity
  • Availability
  • Add to Cart