ENA371178362 Enamine

N-(2-hydroxyethyl)-N-methylmethanesulfonamide

  • MDL number MFCD11934382
  • Cas Number : 87113-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86450
Availability details for
Quantity
  • Availability
  • Add to Cart