ENA371178364 Enamine

2-(1-chloroethyl)-5-(3,4-dimethylphenyl)-1,3,4-oxadiazole

  • MDL number MFCD12188313
  • Cas Number : 1156945-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106509
Availability details for
Quantity
  • Availability
  • Add to Cart