ENA371178372 Enamine

1-(2-methylthiolan-2-yl)methanamine

  • MDL number MFCD22375436
  • Cas Number : 1384665-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93814
Availability details for
Quantity
  • Availability
  • Add to Cart