ENA371178373 Enamine

N-[(morpholin-2-yl)methyl]pyridazin-3-amine

  • MDL number MFCD22375437
  • Cas Number : 1384691-11-5
  • Molecular Weight : 194.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2-morpholinylmethyl)-3-pyridazinamine
Inchi Code 1S/C9H14N4O/c1-2-9(13-12-3-1)11-7-8-6-10-4-5-14-8/h1-3,8,10H,4-7H2,(H,11,13)
Inchi Key KHMVPKJFFVRRFV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-94038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94038
Availability details for
Quantity
  • Availability
  • Add to Cart