ENA371178377 Enamine

3-(1H-imidazol-1-yl)-4-methylpiperidine

  • MDL number MFCD21740663
  • Cas Number : 1384626-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106514
Availability details for
Quantity
  • Availability
  • Add to Cart