ENA371178384 Enamine

3-(2-aminopropan-2-yl)-1,2,4-oxadiazole-5-carboxamide

  • MDL number MFCD22375442
  • Cas Number : 1394041-62-3
  • Molecular Weight : 170.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(1-amino-1-methylethyl)-1,2,4-oxadiazole-5-carboxamide
Inchi Code 1S/C6H10N4O2/c1-6(2,8)5-9-4(3(7)11)12-10-5/h8H2,1-2H3,(H2,7,11)
Inchi Key YYMPVUCUGGRPRH-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-99022

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-99022
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart