ENA371178385 Enamine

tert-butyl N-[2-(1,2,4-oxadiazol-3-yl)propan-2-yl]carbamate

  • MDL number MFCD22375443
  • Cas Number : 1394040-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99753
Availability details for
Quantity
  • Availability
  • Add to Cart