ENA371178388 Enamine

2-(chloromethyl)-2,3-dihydro-1-benzofuran

  • MDL number MFCD11646265
  • Cas Number : 53491-32-0
  • Molecular Weight : 168.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-96359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96359
Availability details for
Quantity
  • Availability
  • Add to Cart