ENA371178403 Enamine

1-(3,3-diethoxypropoxy)-4-fluorobenzene

  • MDL number MFCD18989410
  • Cas Number : 1394042-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98958
Availability details for
Quantity
  • Availability
  • Add to Cart