ENA371178404 Enamine

4-(3,3-diethoxypropoxy)benzonitrile

  • MDL number MFCD18598398
  • Cas Number : 1394040-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98960
Availability details for
Quantity
  • Availability
  • Add to Cart