ENA371178405 Enamine

3-(3,3-diethoxypropoxy)benzonitrile

  • MDL number MFCD19507849
  • Cas Number : 1394041-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98961

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98961
Availability details for
Quantity
  • Availability
  • Add to Cart