ENA371463124 Enamine

4-chloro-2-methyl-5-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD22378588
  • Cas Number : 98273-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105810

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105810
Availability details for
Quantity
  • Availability
  • Add to Cart