ENA371463126 Enamine

4-(2-bromoethoxy)-3-chlorobenzaldehyde

  • MDL number MFCD22378591
  • Cas Number : 1394040-83-5
  • Molecular Weight : 263.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106761
Availability details for
Quantity
  • Availability
  • Add to Cart