ENA371463127 Enamine

2-(4H-1,2,4-triazol-3-yl)phenol

  • MDL number MFCD22378592
  • Cas Number : 91983-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-107034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107034
Availability details for
Quantity
  • Availability
  • Add to Cart