ENA371463131 Enamine

methyl 2-iodo-5-methyl-3-nitrobenzoate

  • MDL number MFCD22378596
  • Cas Number : 854645-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107415
Availability details for
Quantity
  • Availability
  • Add to Cart