ENA371463133 Enamine

tert-butyl 8-formyl-1,2,3,4-tetrahydroquinoline-1-carboxylate

  • MDL number MFCD22378599
  • Cas Number : 444188-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107757
Availability details for
Quantity
  • Availability
  • Add to Cart