ENA371463135 Enamine

[(4-chloro-1H-pyrrol-2-yl)methyl](methyl)amine

  • MDL number MFCD22378601
  • Cas Number : 1394041-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107836
Availability details for
Quantity
  • Availability
  • Add to Cart