ENA371463139 Enamine

1,4-dioxepan-6-amine hydrochloride

  • MDL number MFCD22378607
  • Cas Number : 1394042-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108171
Availability details for
Quantity
  • Availability
  • Add to Cart