ENA371463140 Enamine

(1S)-1-(1-phenyl-1H-pyrazol-4-yl)ethan-1-ol

  • MDL number MFCD22378608
  • Cas Number : 1394051-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108228
Availability details for
Quantity
  • Availability
  • Add to Cart