ENA371463141 Enamine

sodium 2-[(diethylamino)methyl]benzene-1-sulfonate

  • MDL number MFCD22378609
  • Cas Number : 1394040-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108414

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108414
Availability details for
Quantity
  • Availability
  • Add to Cart