ENA371463145 Enamine

2-{[5-(trifluoromethyl)pyridin-2-yl]oxy}ethan-1-amine hydrochloride

  • MDL number MFCD22378613
  • Cas Number : 898271-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108577
Availability details for
Quantity
  • Availability
  • Add to Cart