ENA371463150 Enamine

N-(pyridin-3-yl)piperidine-4-carboxamide dihydrochloride

  • MDL number MFCD22378620
  • Cas Number : 110106-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108653

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108653
Availability details for
Quantity
  • Availability
  • Add to Cart