ENA371463158 Enamine

5-tert-butyl-1,3-oxazole-2-thiol

  • MDL number MFCD22378631
  • Cas Number : 58417-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108842
Availability details for
Quantity
  • Availability
  • Add to Cart