ENA371463159 Enamine

5-benzyloxolan-2-one

  • MDL number MFCD22378632
  • Cas Number : 21175-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30-32
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108845
Availability details for
Quantity
  • Availability
  • Add to Cart